Anti-RBP4 Rabbit Monoclonal Antibody [clone: 003]

Supplier: Sino Biological

80008-R003-50 80008-R003-100
103596-556EA 128.15 USD
103596-556 103660-428
Anti-RBP4 Rabbit Monoclonal Antibody [clone: 003]
Antibodies
Anti-RBP4 Antibody

Type: Primary
Antigen: RBP4
Clonality: Monoclonal
Clone: Clone: 003
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR