Anti-CD155 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80007-RP02-200 80007-RP02-50 80007-RP02-100
103660-426EA 214.31 USD
103660-426 103596-550 103660-424
Anti-CD155 Rabbit Polyclonal Antibody
Antibodies
Anti-CD155 / PVR Antibody

Type: Primary
Antigen: CD155
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR