Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD155 Mouse Monoclonal Antibody [clone: 4F10H8A11]

Supplier: Sino Biological

80007-MM06-200 80007-MM06-100 80007-MM06-50
103660-414EA 214.31 USD InStock
103660-414 103660-412 103596-546
Anti-CD155 Mouse Monoclonal Antibody [clone: 4F10H8A11]
Antibodies
Anti-CD155 / PVR Antibody

Type: Primary
Antigen: CD155
Clonality: Monoclonal
Clone: Clone: 4F10H8A11
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty