Anti-VEGFA Rabbit Polyclonal Antibody

Supplier: Sino Biological
80006-RP01-100 80006-RP01-200 80006-RP01-400
103660-408EA 270.2 USD
103660-408 103660-406 103660-410
Anti-VEGFA Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat VEGF164/VEGFA (Catalog#80006-RNAB; AAA41211.1; Met1-Arg190). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: VEGF164
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR