Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PCSK9 Mouse Monoclonal Antibody [clone: 04]

Supplier: Sino Biological

80005-MM04-50 80005-MM04-100 80005-MM04-200
103596-530EA 106.62 USD InStock
103596-530 103660-862 103660-864
Anti-PCSK9 Mouse Monoclonal Antibody [clone: 04]
Antibodies
Anti-PCSK9 Antibody

Type: Primary
Antigen: PCSK9
Clonality: Monoclonal
Clone: Clone: 04
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty