Anti-PCSK9 Mouse Monoclonal Antibody [clone: 03]

Supplier: Sino Biological
80005-MM03-200 80005-MM03-50 80005-MM03-100
103596-528EA 142.96 USD
103596-528 103660-858 103660-860
Anti-PCSK9 Mouse Monoclonal Antibody [clone: 03]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat PCSK9 (rR PCSK9; Catalog#80005-R08H; NP_954862.2; Met 1-Gln 691). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: PCSK9
Clonality: Monoclonal
Clone: 03
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR