Anti-TIM-1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

80003-RP01-400 80003-RP01-100 80003-RP01-200
103660-382EA 214.31 USD
103660-382 103660-378 103660-380
Anti-TIM-1 Rabbit Polyclonal Antibody
Antibodies
Anti-TIM-1 Antibody

Type: Primary
Antigen: TIM-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR