Anti-Gpt2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80002-RP02-200 80002-RP02-100 80002-RP02-50
103596-518EA 142.96 USD
103596-518 103660-370 103660-368
Anti-Gpt2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat GPT2 (rR GPT2; Catalog#80002-R08B; Met 1-Ser 522; NP_001012057.1). GPT2 specific IgG was purified by Rat GPT2 affinity chromatography.

Type: Primary
Antigen: GPT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR