Anti-Gpt2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
80002-RP01-200 80002-RP01-400 80002-RP01-100
103660-362EA 351.49 USD
103660-362 103660-366 103660-364
Anti-Gpt2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Rat GPT2 (Catalog#80002-R08B; NP_001012057.1; Met1-Ser522). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: GPT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR