Anti-Gpt2 Rabbit Monoclonal Antibody [clone: 033]

Supplier: Sino Biological
80002-R033-50 80002-R033-100
103596-516EA 150.18 USD
103596-516 103660-856
Anti-Gpt2 Rabbit Monoclonal Antibody [clone: 033]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Rat GPT2 (rR GPT2; Catalog#80002-R08B; Met 1-Ser 522; NP_001012057.1).

Type: Primary
Antigen: GPT2
Clonality: Monoclonal
Clone: 033
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR