Anti-FOLR1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

70115-T16-200 70115-T16-100 70115-T16-50
103596-514EA 214.31 USD
103596-514 103596-512 103589-652
Anti-FOLR1 Rabbit Polyclonal Antibody
Antibodies
Anti-FOLR1 Antibody

Type: Primary
Antigen: FOLR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR