Anti-CTLA-4 Rabbit Polyclonal Antibody

Supplier: Sino Biological

70113-RP01-200 70113-RP01-100 70113-RP01-400
103660-350EA 160.46 USD
103660-350 103660-352 103660-354
Anti-CTLA-4 Rabbit Polyclonal Antibody
Antibodies
Anti-CTLA-4 Antibody

Type: Primary
Antigen: CTLA-4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR