Anti-IL17RD Rabbit Polyclonal Antibody

Supplier: Sino Biological
70104-T16-50 70104-T16-200 70104-T16-100
103596-498EA 270.87 USD
103596-498 103596-500 103589-646
Anti-IL17RD Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Canine IL-17RD (Catalog#70104-D08H; XP_541835.3; Ala27-Arg299). IL-17RD specific IgG was purified by Canine IL-17RD affinity chromatography.

Type: Primary
Antigen: IL17RD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR