Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TGFB1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

70087-T52-50 70087-T52-100 70087-T52-200
103589-642EA 106.62 USD InStock
103589-642 103596-490 103596-492
Anti-TGFB1 Rabbit Polyclonal Antibody
Antibodies
Anti-TGF beta 1 Antibody

Type: Primary
Antigen: TGFB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty