Anti-CXCL13 Rabbit Polyclonal Antibody

Supplier: Sino Biological
70057-T16-200 70057-T16-100 70057-T16-50
103596-480EA 270.87 USD
103596-480 103589-638 103596-482
Anti-CXCL13 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Canine BCA-1 / CXCL13 (Catalog#70057-D07E; XP_851619.3; Val23-Asn110). BCA-1 / CXCL13 specific IgG was purified by Canine BCA-1 / CXCL13 affinity chromatography.

Type: Primary
Antigen: BCA-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR