Anti-CXCL13 Mouse Monoclonal Antibody [clone: 13]

Supplier: Sino Biological
70057-MM13-100 70057-MM13-50 70057-MM13-200
103660-326EA 352.03 USD
103660-326 103596-478 103660-324
Anti-CXCL13 Mouse Monoclonal Antibody [clone: 13]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Canine BCA-1/CXCL13 ( Catalog#70057-D07E; XP_851619.3; Val23-Asn110). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: BCA-1
Clonality: Monoclonal
Clone: 13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR