Anti-FGF1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
70039-RP01-100 70039-RP01-200 70039-RP01-400
103660-848EA 270.2 USD
103660-848 103660-846 103660-322
Anti-FGF1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Canine aFGF / FGF1 (Catalog#70039-DNAE; XP_849274.1; Phe16-Asp155). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: FGF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR