Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ANG2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

70034-T16-50 70034-T16-200 70034-T16-100
103589-632EA 106.62 USD InStock
103589-632 103596-468 103596-466
Anti-ANG2 Rabbit Polyclonal Antibody
Antibodies
Anti-Angiopoietin-2 Antibody

Type: Primary
Antigen: ANG2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty