Anti-CA9 Rabbit Polyclonal Antibody

Supplier: Sino Biological
70028-T52-100 70028-T52-200 70028-T52-50
103594-678EA 270.87 USD
103594-678 103589-630 103596-464
Anti-CA9 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Canine Carbonic Anhydrase IX / CA9 (Catalog#70028-D08H; NP_001138646.1; Met1-Leu410). Carbonic Anhydrase IX / CA9 specific IgG was purified by Canine Carbonic Anhydrase IX / CA9 affinity chromatography.

Type: Primary
Antigen: CA9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR