Anti-IL-4 Rabbit Polyclonal Antibody

Supplier: Sino Biological

70021-RP01-200 70021-RP01-400 70021-RP01-100
103660-310EA 160.46 USD
103660-310 103660-312 103660-308
Anti-IL-4 Rabbit Polyclonal Antibody
Antibodies
Anti-IL-4 Antibody

Type: Primary
Antigen: IL-4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR