Anti-IL-5 Rabbit Polyclonal Antibody

Supplier: Sino Biological

70019-RP01-200 70019-RP01-100 70019-RP01-400
103660-302EA 160.46 USD
103660-302 103660-304 103660-306
Anti-IL-5 Rabbit Polyclonal Antibody
Antibodies
Anti-IL-5 Antibody

Type: Primary
Antigen: IL-5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR