Anti-IL2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
70014-T48-50 70014-T48-100 70014-T48-200
103596-460EA 210.04 USD
103596-460 103596-458 103589-624
Anti-IL2 Rabbit Polyclonal Antibody
Antibodies
Anti-IL2 Antibody

Type: Primary
Antigen: IL2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR