Anti-EGF Rabbit Polyclonal Antibody

Supplier: Sino Biological
70013-RP01-400 70013-RP01-200 70013-RP01-100
103660-286EA 562.97 USD
103660-286 103660-284 103660-282
Anti-EGF Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Canine EGF / Epidermal Growth Factor (Catalog#70013-D07E; Q9BEA0; Asn973-Arg1024). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR