Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EGF Mouse Monoclonal Antibody [clone: 07]

Supplier: Sino Biological

70013-MM07-100 70013-MM07-200 70013-MM07-50
103660-278EA 160.46 USD InStock
103660-278 103660-280 103596-454
Anti-EGF Mouse Monoclonal Antibody [clone: 07]
Antibodies
Anti-EGF Antibody

Type: Primary
Antigen: EGF
Clonality: Monoclonal
Clone: Clone: 07
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Dog

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty