Anti-c-MET Rabbit Polyclonal Antibody

Supplier: Sino Biological

70008-RP02-100 70008-RP02-200 70008-RP02-50
103660-274EA 160.46 USD
103660-274 103660-276 103596-452
Anti-c-MET Rabbit Polyclonal Antibody
Antibodies
Anti-c-MET Antibody

Type: Primary
Antigen: c-MET
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR