Anti-MET Rabbit Monoclonal Antibody [clone: 345]

Supplier: Sino Biological
70008-R345-50 70008-R345-100
103596-450EA 150.18 USD
103596-450 103660-266
Anti-MET Rabbit Monoclonal Antibody [clone: 345]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Canine HGFR / c-MET (Catalog#70008-D08H; NP_001002963.1; Met1-Leu935).

Type: Primary
Antigen: c-MET
Clonality: Monoclonal
Clone: 345
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR