Anti-RBP4 Rabbit Polyclonal Antibody

Supplier: Sino Biological
70006-T48-200 70006-T48-50 70006-T48-100
103596-444EA 352.03 USD
103596-444 103589-622 103596-442
Anti-RBP4 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Canine RBP4 (Catalog#70006-D08H; F1PQI4; Met1-Leu201). RBP4 specific IgG was purified by Canine RBP4 affinity chromatography.

Type: Primary
Antigen: RBP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR