Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFA Rabbit Polyclonal Antibody

Supplier: Sino Biological

70003-RP02-200 70003-RP02-100 70003-RP02-50
103660-254EA 214.31 USD InStock
103660-254 103660-252 103596-434
Anti-TNFA Rabbit Polyclonal Antibody
Antibodies
Anti-TNF-alpha Antibody

Type: Primary
Antigen: TNFA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty