Anti-TNF Rabbit Polyclonal Antibody

Supplier: Sino Biological
70003-RP01-400 70003-RP01-100 70003-RP01-200
103660-250EA 562.97 USD
103660-250 103660-248 103660-246
Anti-TNF Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Canine TNF-alpha (rCanine TNF-alpha; Catalog#70003-DNAE; NP_001003244.4; Val 77-Leu 233). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: TNFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR