Anti-HAVCR1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
70001-RP02-50 70001-RP02-100 70001-RP02-200
103660-242EA 352.03 USD
103660-242 103660-240 103596-430
Anti-HAVCR1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Canine TIMD1/HAVCR1 extracellular domain (rCanine HAVCR1; Catalog#70001-D07H; XP_854888.1; Tyr 21-Ser 160). TIMD1 specific IgG was purified by canine TIMD1 affinity chromatography.

Type: Primary
Antigen: TIM-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR