Anti-HAVCR1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
70001-RP01-100 70001-RP01-200 70001-RP01-400
103660-238EA 562.97 USD
103660-238 103659-826 103659-828
Anti-HAVCR1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Canine HAVCR1 extracellular domain (rCanine TIMD-1 / HAVCR1; Catalog#70001-D07H; XP_854888.1; Tyr 21-Ser 160). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: TIM-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR