Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HLA-DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: XM01]

Supplier: Sino Biological

68038-XM01-P-25 68038-XM01-P-100
103631-524EA 53.85 USD InStock
103631-524 103632-392
Anti-HLA-DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: XM01]
Antibodies
Anti-HLA-DR Antibody (PE)

Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: Clone: XM01
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty