Anti-CD45RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: XM01]

Supplier: Sino Biological

68036-XM01-P-100 68036-XM01-P-25
103632-388EA 53.85 USD
103632-388 103631-520
Anti-CD45RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: XM01]
Antibodies
Anti-CD45RA Antibody (PE)

Type: Primary
Antigen: CD45RA
Clonality: Monoclonal
Clone: Clone: XM01
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR