Anti-CD45RA Mouse Monoclonal Antibody [clone: XM01] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
68036-XM01-F-25 68036-XM01-F-100
103631-518EA 30.3 USD
103631-518 103632-546
Anti-CD45RA Mouse Monoclonal Antibody [clone: XM01] FITC (Fluorescein Isothiocyanate)
Antibodies
The monoclonal antibody was purified by affinity chromatography. Then it was conjugated with FITC under optimum conditions, and the unreacted FITC was removed.

Type: Primary
Antigen: CD45RA
Clonality: Monoclonal
Clone: Clone: XM01
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR