Anti-Hemagglutinin/ HA Neutralizing Antibody [clone: H011]

Supplier: Sino Biological
68031-H011-500 68031-H011-200
103660-234EA 352.8 USD
103660-234 103660-232
Anti-Hemagglutinin/ HA Neutralizing Antibody [clone: H011]
Antibodies
Influenza H5N1 Hemagglutinin.

Type: Primary
Antigen: Hemagglutinin/ HA Neutralizing
Clonality:
Clone: H011
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity: H5N1 (A/Hubei/1/2010)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR