Anti-M13 Rabbit Polyclonal Antibody

Supplier: Sino Biological

68007-T32-100 68007-T32-200 68007-T32-50
103596-426EA 160.46 USD
103596-426 103594-684 103589-614
Anti-M13 Rabbit Polyclonal Antibody
Antibodies
Anti-M13 Antibody

Type: Primary
Antigen: M13
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR