Anti-ALB Rabbit Polyclonal Antibody

Supplier: Sino Biological
68001-T16-50 68001-T16-100
103596-424EA 279.29 USD
103596-424 103589-612
Anti-ALB Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Human Serum Albumin / HSA / HAS / ALB. Human Serum Albumin / HSA / HAS / ALB specific IgG was purified by Human Serum Albumin / HSA / HAS / ALB affinity chromatography.

Type: Primary
Antigen: HSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR