Anti-ALB Rabbit Monoclonal Antibody [clone: 101] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
68001-R101-F-25 68001-R101-F-100
103631-516EA 90.9 USD
103631-516 103632-544
Anti-ALB Rabbit Monoclonal Antibody [clone: 101] FITC (Fluorescein Isothiocyanate)
Antibodies
This antibody was obtained from a rabbit immunized with purified Human Serum Albumin, and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: HSA
Clonality: Monoclonal
Clone: Clone: 101
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR