Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSA Mouse Monoclonal Antibody [clone: 06]

Supplier: Sino Biological

68001-MM06-50 68001-MM06-200 68001-MM06-100
103596-420EA 106.62 USD InStock
103596-420 103660-226 103660-224
Anti-HSA Mouse Monoclonal Antibody [clone: 06]
Antibodies
Anti-Serum Albumin Antibody

Type: Primary
Antigen: HSA
Clonality: Monoclonal
Clone: Clone: 06
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty