Anti-IL1B Rabbit Polyclonal Antibody

Supplier: Sino Biological
67002-T16-200 67002-T16-50 67002-T16-100
103596-418EA 352.03 USD
103596-418 103596-416 103589-610
Anti-IL1B Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Feline IL-1 beta (Catalog#67002-UNAE; NP_001070882.1; Ala116-Ser267). IL-1 beta specific IgG was purified by Feline IL-1 beta affinity chromatography.

Type: Primary
Antigen: IL-1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR