Anti-CD38 Mouse Monoclonal Antibody [clone: 01]

Supplier: Sino Biological
65003-MM01-200 65003-MM01-100 65003-MM01-50
103596-410EA 142.96 USD
103596-410 103660-216 103660-214
Anti-CD38 Mouse Monoclonal Antibody [clone: 01]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rabbit CD38 (Catalog#65003-T08H; Q9MZ03; Leu43-Met298).

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: 01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR