Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EFNB2A Rabbit Polyclonal Antibody

Supplier: Sino Biological

63002-T16-50 63002-T16-200 63002-T16-100
103589-470EA 106.62 USD InStock
103589-470 103596-408 103596-406
Anti-EFNB2A Rabbit Polyclonal Antibody
Antibodies
Anti-EFNB2A Antibody

Type: Primary
Antigen: EFNB2A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Zebrafish

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty