Anti-IL-6 Rabbit Polyclonal Antibody

Supplier: Sino Biological

62006-T16-200 62006-T16-50 62006-T16-100
103596-404EA 214.31 USD
103596-404 103589-468 103596-402
Anti-IL-6 Rabbit Polyclonal Antibody
Antibodies
Anti-IL-6 Antibody

Type: Primary
Antigen: IL-6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR