Anti-MS4A1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
60004-RP02-100 60004-RP02-50 60004-RP02-200
103660-834EA 352.03 USD
103660-834 103596-388 103660-200
Anti-MS4A1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Ferret CD20 / MS4A1 (rF CD20 / MS4A1; Catalog#60004-F08H; Glu 213-Pro 297). CD20 / MS4A1 specific IgG was purified by Ferret CD20 / MS4A1 affinity chromatography.

Type: Primary
Antigen: CD20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Ferret
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR