Anti-CD4 Rabbit Polyclonal Antibody

Supplier: Sino Biological
60003-RP02-200 60003-RP02-100 60003-RP02-50
103660-190EA 352.03 USD
103660-190 103596-384 103660-188
Anti-CD4 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Ferret CD4 (rF CD4; Catalog#60003-F08H; ABS50090.1; Met 1-Leu 401). CD4 specific IgG was purified by Ferret CD4 affinity chromatography.

Type: Primary
Antigen: Anti-CD4 Antibody
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Ferret
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR