Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Anti-CD4 Antibody Mouse Monoclonal Antibody [clone: 08]

Supplier: Sino Biological

60003-MM08-1 60003-MM08-200 60003-MM08-50 60003-MM08-100
103589-604EA 958.46 USD InStock
103589-604 103660-180 103596-378 103660-178
Anti-Anti-CD4 Antibody Mouse Monoclonal Antibody [clone: 08]
Antibodies
Anti-CD4 Antibody

Type: Primary
Antigen: Anti-CD4 Antibody
Clonality: Monoclonal
Clone: Clone: 08
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Ferret

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty