Anti-CD4 Mouse Monoclonal Antibody [clone: 02] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
60003-MM02-F-25 60003-MM02-F-100
103632-540EA 181.8 USD
103632-540 103630-208
Anti-CD4 Mouse Monoclonal Antibody [clone: 02] FITC (Fluorescein Isothiocyanate)
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ferret CD4 (rF CD4; Catalog#60003-F08H; ABS50090.1; Met 1-Leu 401) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: Clone: 02
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Ferret
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR