Anti-CD4 Mouse Monoclonal Antibody [clone: 02]

Supplier: Sino Biological
60003-MM02-100 60003-MM02-50 60003-MM02-1
103660-176EA 301.55 USD
103660-176 103589-602 103596-376
Anti-CD4 Mouse Monoclonal Antibody [clone: 02]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ferret CD4 (rF CD4; Catalog#60003-F08H; ABS50090.1; Met 1-Leu 401). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: Anti-CD4 Antibody
Clonality: Monoclonal
Clone: 02
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Ferret
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR