Anti-TNF Rabbit Monoclonal Antibody [clone: 030]

Supplier: Sino Biological
60002-R030-100 60002-R030-50
103596-368EA 150.18 USD
103596-368 103660-166
Anti-TNF Rabbit Monoclonal Antibody [clone: 030]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Ferret TNF-alpha / TNFA (Catalog#60002-FNAE; A3FBF1; Val77-Leu233).

Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: 030
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Ferret
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR