Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFA Mouse Monoclonal Antibody [clone: 11]

Supplier: Sino Biological

60002-MM11-200 60002-MM11-1 60002-MM11-50 60002-MM11-100
103660-164EA 214.31 USD InStock
103660-164 103589-596 103596-366 103660-162
Anti-TNFA Mouse Monoclonal Antibody [clone: 11]
Antibodies
Anti-TNF-alpha Antibody

Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: Clone: 11
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Ferret

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty